missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human DNA ligase 1 Protein
A DNA sequence encoding the Homo sapiens (Human) DNA ligase 1, was expressed in the hosts and tags indicated.
160.00 € - 2385.00 €
Specifications
Accession Number | NP_000225.1 |
---|---|
Gene ID (Entrez) | 3978 |
Name | DNA ligase 1 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16072360
|
enQuireBio™
QP6308-EC-100UG |
100 μg |
530.00 €
100µg |
Estimated Shipment: 25-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16092360
|
enQuireBio™
QP6308-EC-1MG |
1 mg |
2385.00 €
1mg |
Estimated Shipment: 25-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16082360
|
enQuireBio™
QP6308-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16002370
|
enQuireBio™
QP6308-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16012370
|
enQuireBio™
QP6308-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16022370
|
enQuireBio™
QP6308-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_000225.1 | |
DNA ligase 1 Protein | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |
3978 | |
Research Use Only | |
LIG1 | |
Recombinant Protein | |
E. coli | |
MQRSIMSFFH PKKEGKAKKP EKEASNSSRE TEPPPKAALK EWNGVVSESD SPVKRPGRKA ARVLGSEGEE EDEALSPAKG QKPALDCSQV SPPRPATSPE NNASLSDTSP MDSSPSGIPK RRTARKQLPK RTIQEVLEEQ SEDEDREAKR KKEEEEEETP KESLTEAEVA TEKEGEDGDQ PTTPPKPLKT SKAETPTESV SEPEVATKQE LQEEEEQTKP PRRAPKTLSS FFTPRKPAVK KEVKEEEPGA PGKEGAAEGP LDPSGYNPAK NNYHPVEDAC WKPGQKVPYL AVARTFEKIE EVSARLRMVE TLSNLLRSVV ALSPPDLLPV LYLSLNHLGP PQQGLELGVG DGVLLKAVAQ ATGRQLESVR AEAAEKGDVG LVAENSRSTQ RLMLPPPPLT ASGVFSKFRD IARLTGSAST AKKIDIIKGL FVACRHSEAR FIARSLSGRL RLGLAEQSVL AALSQAVSLT PPGQEFPPAM VDAGKGKTAE ARKTWLEEQG MILKQTFCEV PDLDRIIPVL LEHGLERLPE HCKLSPGIPL KPMLAHPTRG ISEVLKRFEE AAFTCEYKYD GQRAQIHALE GGEVKIFSRN QEDNTGKYPD IISRIPKIKL PSVTSFILDT EAVAWDREKK QIQPFQVLTT RKRKEVDASE IQVQVCLYAF DLIYLNGESL VREPLSRRRQ LLRENFVETE GEFVFATSLD TKDIEQIAEF LEQSVKDSCE GLMVKTLDVD ATYEIAKRSH NWLKLKKDYL DGVGDTLDLV VIGAYLGRGK RAGRYGGFLL ASYDEDSEEL QAICKLGTGF SDEELEEHHQ SLKALVLPSP RPYVRIDGAV IPDHWLDPSA VWEVKCADLS LSPIYPAARG LVDSDKGISL RFPRFIRVRE DKQPEQATTS AQVACLYRKQ SQIQNQQGED | |
Greater than 90% as determined by SDS-PAGE. |