missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Cytokeratin 7 Protein
A DNA sequence encoding the Homo sapiens (Human) Cytokeratin 7, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6283-ec-500ug
Additional Details : Weight : 0.01000kg
Specifications
NP_005547.3 | |
Cytokeratin 7 Protein | |
Research Use Only | |
KRT7 | |
Recombinant Protein | |
E. coli | |
SIHFSSPVFT SRSAAFSGRG AQVRLSSARP GGLGSSSLYG LGASRPRVAV RSAYGGPVGA GIREVTINQS LLAPLRLDAD PSLQRVRQEE SEQIKTLNNK FASFIDKVRF LEQQNKLLET KWTLLQEQKS AKSSRLPDIF EAQIAGLRGQ LEALQVDGGR LEAELRSMQD VVEDFKNKYE DEINHRTAAE NEFVVLKKDV DAAYMSKVEL EAKVDALNDE INFLRTLNET ELTELQSQIS DTSVVLSMDN SRSLDLDGII AEVKAQYEEM AKCSRAEAEA WYQTKFETLQ AQAGKHGDDL RNTRNEISEM NRAIQRLQAE IDNIKNQRAK LEAAIAEAEE RGELALKDAR AKQEELEAAL QRGKQDMARQ LREYQELMSV KLALDIEIAT YRKLLEGEES RLAGDGVGAV NISVMNSTGG SSSGGGIGLT LGGTMGSNAL SFSSSAGPGL LKAYSIRTAS ASRRSARD | |
Greater than 90% as determined by SDS-PAGE. |
3855 | |
500 μg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |