missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Cytokeratin 7 Protein

A DNA sequence encoding the Homo sapiens (Human) Cytokeratin 7, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6283-ec-10ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16062170

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

Specifications

Specifications

NP_005547.3
Cytokeratin 7 Protein
Research Use Only
KRT7
Recombinant Protein
E. coli
SIHFSSPVFT SRSAAFSGRG AQVRLSSARP GGLGSSSLYG LGASRPRVAV RSAYGGPVGA GIREVTINQS LLAPLRLDAD PSLQRVRQEE SEQIKTLNNK FASFIDKVRF LEQQNKLLET KWTLLQEQKS AKSSRLPDIF EAQIAGLRGQ LEALQVDGGR LEAELRSMQD VVEDFKNKYE DEINHRTAAE NEFVVLKKDV DAAYMSKVEL EAKVDALNDE INFLRTLNET ELTELQSQIS DTSVVLSMDN SRSLDLDGII AEVKAQYEEM AKCSRAEAEA WYQTKFETLQ AQAGKHGDDL RNTRNEISEM NRAIQRLQAE IDNIKNQRAK LEAAIAEAEE RGELALKDAR AKQEELEAAL QRGKQDMARQ LREYQELMSV KLALDIEIAT YRKLLEGEES RLAGDGVGAV NISVMNSTGG SSSGGGIGLT LGGTMGSNAL SFSSSAGPGL LKAYSIRTAS ASRRSARD
Greater than 90% as determined by SDS-PAGE.
3855
10 μg
Not determined.
Full Length
Human
His-SUMO
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions