missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human CXXC-type zinc finger protein 1
A DNA sequence encoding the Homo sapiens (Human) CXXC-type zinc finger protein 4, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8141-ec-1mg
Additional Details : Weight : 0.01000kg
Specifications
CXXC-type zinc finger protein 1 | |
Research Use Only | |
CXXC4 | |
Recombinant Protein | |
E. coli | |
MHHRNDSQRL GKAGCPPEPS LQMANTNFLS TLSPEHCRPL AGECMNKLKC GAAEAEIMNL PERVGTFSAI PALGGISLPP GVIVMTALHS PAAASAAVTD SAFQIANLAD CPQNHSSSSS SSSGGAGGAN PAKKKRKRCG VCVPCKRLIN CGVCSSCRNR KTGHQICKFR KCEELKKKPG TSLERTPVPS AEAFRWFF | |
Greater than 80% as determined by SDS-PAGE. |
1 mg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |