missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Recombinant Human CXCL2/GRO beta/MIP-2/CINC-3 Protein
Shop All Bio Techne Products
Click to view available options
Quantity:
1 mg
10 μg
100 μg
500 μg
Unit Size:
100µg
10µg
1mg
500µg
Description
CXCL2 Protein is fully biologically active when compared to standard. The biological activity determined by a chemotaxis bioassay using human CXCR2 transfected human 293 cells is in a concentration range of 10-100 ng/mL.
Specifications
Specifications
| Accession Number | P19875, P19875, P19875 |
| Concentration | LYOPH |
| For Use With (Application) | PAGE, Bioactivity |
| Formulation | Lyophilized from a 0.2 um filtered concentrated solution in 20 mM PB, pH 7.4, 50 mM NaCl. |
| Gene ID (Entrez) | 2920 |
| Molecular Weight (g/mol) | 7.9 kDa |
| Quantity | 100 μg |
| Storage Requirements | Store at -20°C to -70°C as supplied. After reconstitution, store at 2°C to 8°C for 1 month and at -20°C to -70°C for long term storage. Avoid repeated freeze-thaw cycles. |
| Endotoxin Concentration | Less than 1 EU/ug of CXCL2/GRO beta/MIP-2/CINC-3 as determined by LAL method. |
| Gene Alias | chemokine (C-X-C motif) ligand 2, CINC-2a, CINC-3/CXCL2, C-X-C Motif Chemokine 2, GRO2, GRO2 oncogene, GROB, Gro-beta, Growth-regulated protein beta, Macrophage Inflammatory Protein, Macrophage Inflammatory Protein 2, Macrophage inflammatory protein 2-alpha, melanoma growth stimulatory activity beta, MGSA beta, MGSA-b, MIP2, MIP-2/CXCL2, MIP-2a, MIP2A, MIP2-alpha, SCYB2 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction