missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
CXCL17 protein is fully biologically active when compared to standard. The ED50 as determined by its ability to induce VEGF expression using murine endothelial cells is less than 5.0 μg/mL, corresponding to a specific activity of >200 IU/mg.
Specifications
Specifications
| Accession Number | Q6UXB2, Q6UXB2, Q6UXB2 |
| For Use With (Application) | PAGE, Bioactivity |
| Formulation | Lyophilized from a 0.2 um filtered concentrated solution in PBS, pH 7.4, with 3 % Trehalose. |
| Gene ID (Entrez) | 284340 |
| Molecular Weight (g/mol) | M.W. (Theoretical): 11.5 kDa |
| Quantity | 500 μg |
| Storage Requirements | Store at -20°C to -70°C as supplied. After reconstitution, store at 2°C to 8°C for 1 month and at -20°C to -70°C for long term storage. Avoid repeated freeze-thaw cycles. |
| Endotoxin Concentration | Less than 0.1 EU/μg of CXCL17/VCC-1 as determined by LAL method. |
| Gene Alias | Chemokine (C-X-C Motif) Ligand 17, C-X-C Motif Chemokine 17, Dcip1, Dendritic Cell And Monocyte Chemokine-Like Protein, DMC, UNQ473, VCC1, VCC-1, VCC-1/CXCL17, VEGF Coregulated Chemokine 1, VEGF Co-Regulated Chemokine 1 |
| Gene Symbol | CXCL17 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?