missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Complement component C8 gamma chain Protein

Product Code. 15997399
Change view
Click to view available options
Quantity:
1 mg
10 μg
100 μg
200 μg
50 μg
500 μg
Unit Size:
100µg
10µg
1mg
200µg
500µg
50µg
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15997399 100 μg 100µg
15907409 10 μg 10µg
15947409 50 μg 50µg
15927409 200 μg 200µg
15937409 500 μg 500µg
15917409 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 missing translation for 'options'
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 15997399 missing translation for 'mfr' enQuireBio™ missing translation for 'supplierNo' QP5739ec100ug

Please to purchase this item. Need a web account? Register with us today!

Denna artikel kan inte returneras. Se returpolicy

A DNA sequence encoding the Homo sapiens (Human) Complement component C8 gamma chain, was expressed in the hosts and tags indicated.

Specifikationer

Accession Number NP_000597.2
Gene ID (Entrez) 733
Name Complement component C8 gamma chain Protein
Quantity 100 μg
Regulatory Status Research Use Only
Endotoxin Concentration Not determined.
Gene Symbol C8G
Protein Length Full Length
Product Type Recombinant Protein
Cross Reactivity Human
Species E. coli
Protein Tag His-SUMO
Sequence QKPQRPRRPA SPISTIQPKA NFDAQQFAGT WLLVAVGSAC RFLQEQGHRA EATTLHVAPQ GTAMAVSTFR KLDGICWQVR QLYGDTGVLG RFLLQARDAR GAVHVVVAET DYQSFAVLYL ERAGQLSVKL YARSLPVSDS VLSGFEQRVQ EAHLTEDQIF YFPKYGFCEA ADQFHVLDEV RR
Buffer Tris-based buffer, 50% glycerol
Purity or Quality Grade Greater than 90% as determined by SDS-PAGE.
Visa mer Visa mindre
Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.