missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Complement C4-B Protein
A DNA sequence encoding the Homo sapiens (Human) Complement C4-B, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6998-ye-50ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001002029.3 | |
Complement C4-B Protein | |
Research Use Only | |
C4B | |
Recombinant Protein | |
Yeast | |
EAPKVVEEQE SRVHYTVCIW RNGKVGLSGM AIADVTLLSG FHALRADLEK LTSLSDRYVS HFETEGPHVL LYFDSVPTSR ECVGFEAVQE VPVGLVQPAS ATLYDYYNPE RRCSVFYGAP SKSRLLATLC SAEVCQCAEG KCPRQRRALE RGLQDEDGYR MKFACYYPRV EYGFQVKVLR EDSRAAFRLF ETKITQVLHF TKDVKAAANQ MRNFLVRASC RLRLEPGKEY LIMGLDGATY DLEGHPQYLL DSNSWIEEMP SERLCRSTRQ RAACAQLNDF LQEYGTQGCQ V | |
Greater than 90% as determined by SDS-PAGE. |
100293534 | |
50 μg | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |