missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Claudin-6 Protein

A DNA sequence encoding the Homo sapiens(Human) Claudin-6, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP9973-iv-10ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16009121

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

Specifications

Specifications

NP_067018.2
Claudin-6 Protein
Research Use Only
CLDN6
Recombinant Protein
E. coli
MASAGMQILG VVLTLLGWVN GLVSCALPMW KVTAFIGNSI VVAQVVWEGL WMSCVVQSTG QMQCKVYDSL LALPQDLQAA RA
Greater than 85% as determined by SDS-PAGE.
9074
10 μg
Not determined.
Partial (see sequence information for more details).
Human
His-SUMO
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions