missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human CHCHD4 Protein

A DNA sequence encoding the Homo sapiens (Human) CHCHD4, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7968-ec-10ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16061891

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

Specifications

Specifications

NP_001091972.1
CHCHD4 Protein
Research Use Only
CHCHD4
Recombinant Protein
E. coli
MSYCRQEGKD RIIFVTKEDH ETPSSAELVA DDPNDPYEEH GLILPNGNIN WNCPCLGGMA SGPCGEQFKS AFSCFHYSTE EIKGSDCVDQ FRAMQECMQK YPDLYPQEDE DEEEEREKKP AEQAEETAPI EATATKEEEG SS
Greater than 90% as determined by SDS-PAGE.
131474
10 μg
Not determined.
Full Length
Human
His-SUMO
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions