missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Cell death activator CIDE-3 Protein
A DNA sequence encoding the Homo sapiens (Human) Cell death activator CIDE-3, was expressed in the hosts and tags indicated.
160.00 € - 2385.00 €
Specifications
Accession Number | NP_001186480.1 |
---|---|
Gene ID (Entrez) | 63924 |
Name | Cell death activator CIDE-3 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16071541
|
enQuireBio™
QP7912-EC-100UG |
100 μg |
530.00 €
100µg |
Estimated Shipment: 24-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16091541
|
enQuireBio™
QP7912-EC-1MG |
1 mg |
2385.00 €
1mg |
Estimated Shipment: 24-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16081541
|
enQuireBio™
QP7912-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16001551
|
enQuireBio™
QP7912-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16011551
|
enQuireBio™
QP7912-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16021551
|
enQuireBio™
QP7912-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_001186480.1 | |
Cell death activator CIDE-3 Protein | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |
63924 | |
Research Use Only | |
CIDEC | |
Recombinant Protein | |
E. coli | |
MEYAMKSLSL LYPKSLSRHV SVRTSVVTQQ LLSEPSPKAP RARPCRVSTA DRSVRKGIMA YSLEDLLLKV RDTLMLADKP FFLVLEEDGT TVETEEYFQA LAGDTVFMVL QKGQKWQPPS EQGTRHPLSL SHKPAKKIDV ARVTFDLYKL NPQDFIGCLN VKATFYDTYS LSYDLHCCGA KRIMKEAFRW ALFSMQATGH VLLGTSCYLQ QLLDATEEGQ PPKGKASSLI PTCLKILQ | |
Greater than 80% as determined by SDS-PAGE. |