missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human CD40L / CD154 / TNFSF5 Protein
A DNA sequence encoding the Homo sapiens (Human) CD40L / CD154 / TNFSF5, was expressed in the hosts and tags indicated.
202.00 € - 2155.00 €
Specifications
Accession Number | NP_000065.1 |
---|---|
Gene ID (Entrez) | 959 |
Name | CD40L / CD154 / TNFSF5 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16037531
|
enQuireBio™
QP9047-YE-100UG |
100 μg |
645.00 €
100µg |
Estimated Shipment: 09-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16077531
|
enQuireBio™
QP9047-YE-500UG |
500 μg |
1740.00 €
500µg |
Estimated Shipment: 09-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16047531
|
enQuireBio™
QP9047-YE-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16067531
|
enQuireBio™
QP9047-YE-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16087531
|
enQuireBio™
QP9047-YE-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16057531
|
enQuireBio™
QP9047-YE-1MG |
1 mg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_000065.1 | |
CD40L / CD154 / TNFSF5 Protein | |
Not determined. | |
Extracellular Domain | |
Human | |
His | |
Tris-based buffer, 50% glycerol |
959 | |
Research Use Only | |
CD40LG | |
Recombinant Protein | |
Yeast | |
HRRLDKIEDE RNLHEDFVFM KTIQRCNTGE RSLSLLNCEE IKSQFEGFVK DIMLNKEETK KENSFEMQKG DQNPQIAAHV ISEASSKTTS VLQWAEKGYY TMSNNLVTLE NGKQLTVKRQ GLYYIYAQVT FCSNREASSQ APFIASLCLK SPGRFERILL RAANTHSSAK PCGQQSIHLG GVFELQPGAS VFVNVTDPSQ VSHGTGFTSF GLLKL | |
Greater than 90% as determined by SDS-PAGE. |