missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human CCDC43 Protein
Specifications
Specifications
| Name | CCDC43 Protein |
| Quantity | 2 μg |
| Regulatory Status | Research Use Only |
| Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
| Product Type | Recombinant Protein |
| Cross Reactivity | Human |
| Species | E. coli |
| Protein Tag | His |
| Sequence | MGSSHHHHHH SSGLVPRGSH MGSMAAPSEV AAIAPGEGDG GGGGFGSWLD GRLEALGVDR AVYGAYILGI LQEEEEEEKL DALQGILSAF LEEDSLLNIC KEIVERWSET QNVVTKVKKE DEVQAIATLI EKQAQIVVKP RMVSEEEKQR KAALLAQYAD VTDEEDEADE KDDSGATTMN IGSDKLLFRN TNVEDVLNAR KLERDSLRDE SQRKKEQDKL QRERDKLAKQ ERKEKEKKRT QRGERKR |
| Buffer | The CCDC43 solution (0.25 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 0.15M NaCl, 20% glycerol and 1mM DTT. |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?