missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human CCDC43 Protein

Product Code. 15906738
Change view
Click to view available options
Quantity:
1 mg
10 μg
2 μg
Unit Size:
10µg
1mg
2µg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15906738 10 μg 10µg
15926738 2 μg 2µg
15916738 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15906738 Supplier enQuireBio™ Supplier No. QP1129010ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

A cDNA sequence encoding the CCDC43 was constructed and used to recombinantly synthesize the protein.

Specifications

Name CCDC43 Protein
Quantity 10 μg
Regulatory Status Research Use Only
Endotoxin Concentration Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Product Type Recombinant Protein
Cross Reactivity Human
Species E. coli
Protein Tag His
Sequence MGSSHHHHHH SSGLVPRGSH MGSMAAPSEV AAIAPGEGDG GGGGFGSWLD GRLEALGVDR AVYGAYILGI LQEEEEEEKL DALQGILSAF LEEDSLLNIC KEIVERWSET QNVVTKVKKE DEVQAIATLI EKQAQIVVKP RMVSEEEKQR KAALLAQYAD VTDEEDEADE KDDSGATTMN IGSDKLLFRN TNVEDVLNAR KLERDSLRDE SQRKKEQDKL QRERDKLAKQ ERKEKEKKRT QRGERKR
Buffer The CCDC43 solution (0.25 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 0.15M NaCl, 20% glycerol and 1mM DTT.
Purity or Quality Grade Greater than 85% as determined by SDS-PAGE.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.