missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Cardiac phospholamban Protein
A DNA sequence encoding the Homo sapiens (Human) Cardiac phospholamban, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP9335-ye-10ug
Additional Details : Weight : 0.01000kg
Specifications
NP_002658.1 | |
Cardiac phospholamban Protein | |
Research Use Only | |
P26678 | |
Recombinant Protein | |
Yeast | |
MEKVQYLTRS AIRRASTIEM PQQARQKLQN LFINFCLILI CLLLICIIVM LL | |
Greater than 90% as determined by SDS-PAGE. |
5350 | |
10 μg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |