missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Beta-catenin-interacting protein 1
A DNA sequence encoding the Homo sapiens (Human) Beta-catenin-interacting protein 1, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8120-ec-200ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001012329.1 | |
Beta-catenin-interacting protein 1 | |
Research Use Only | |
CTNNBIP1 | |
Recombinant Protein | |
E. coli | |
MNREGAPGKS PEEMYIQQKV RVLLMLRKMG SNLTASEEEF LRTYAGVVNS QLSQLPPHSI DQGAEDVVMA FSRSETEDRR Q | |
Greater than 80% as determined by SDS-PAGE. |
56998 | |
200 μg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |