missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Human BDNF Protein

Product Code. 18748166
Click to view available options
Quantity:
1 mg
10 μg
250 μg
50 μg
Unit Size:
10µg
1mg
250µg
50µg
This item is not returnable. View return policy

Product Code. 18748166

Brand: R&D Systems™ 11166BD01M

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Measured in a cell proliferation assay using BaF mouse pro-B cells transfected with TrkB. The ED50 for this effect is 0.200-2.00 ng/mL.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P23560.1
For Use With (Application) Bioactivity
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose.
Gene ID (Entrez) 627
Molecular Weight (g/mol) M.W.-observed: 11-15 kDa, under reducing conditions., M.W.-theroretical: 13.5 kDa
Quantity 1 mg
Source Chinese Hamster Ovary cell line, CHO-derived human BDNF protein His129-Arg247
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution.
Endotoxin Concentration <0.10 EU / 1 μg of the protein by the LAL method.
Gene Alias abrineurin, ANON2, brain-derived neurotrophic factor, BULN2, MGC34632, neurotrophin
Conjugate Unconjugated
Reconstitution Reconstitute at 100-500 μg/mL in PBS.
Purity or Quality Grade >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining.
Protein BDNF
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.