missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Baculoviral IAP repeat-containing protein 1
A DNA sequence encoding the Homo sapiens (Human) Baculoviral IAP repeat-containing protein 1, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7547-ec-200ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001333799.1 | |
Baculoviral IAP repeat-containing protein 1 | |
Research Use Only | |
NAIP | |
Recombinant Protein | |
E. coli | |
EAKRLKTFVT YEPYSSWIPQ EMAAAGFYFT GVKSGIQCFC CSLILFGAGL TRLPIEDHKR FHPDCGFLLN KDVGNIAKYD IRVKNLKSRL RGGKMRYQEE EARLASFRNW PFYVQGISPC VLSEAGFVFT GKQDTVQCFS CGGCLGNWEE GDDPWKEHAK WFPKCEFLRS KKSSEEITQY IQSYKGFVDI TGEHFVNSWV QRELPMASAY CNDSIFAYEE LRLDSFKDWP RESAVGVAAL AKAGLFYTGI KDIVQCFSCG GCLEKWQEGD DPLDDHTRCF PNCPFL | |
Greater than 90% as determined by SDS-PAGE. |
4671 | |
200 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |