missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human B9 domain-containing protein 1
A DNA sequence encoding the Homo sapiens (Human) B9 domain-containing protein 2, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8040-ec-200ug
Additional Details : Weight : 0.01000kg
Specifications
NP_085055.2 | |
B9 domain-containing protein 1 | |
Research Use Only | |
B9D2 | |
Recombinant Protein | |
E. coli | |
MAEVHVIGQI IGASGFSESS LFCKWGIHTG AAWKLLSGVR EGQTQVDTPQ IGDMAYWSHP IDLHFATKGL QGWPRLHFQV WSQDSFGRCQ LAGYGFCHVP SSPGTHQLAC PTWRPLGSWR EQLARAFVGG GPQLLHGDTI YSGADRYRLH TAAGGTVHLE IGLLLRNFDR YGVEC | |
Greater than 80% as determined by SDS-PAGE. |
80776 | |
200 μg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |