missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Appetite-regulating hormone Protein
A DNA sequence encoding the Homo sapiens (Human) Appetite-regulating hormone, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8104-ec-50ug
This item is not returnable.
View return policy
Specifications
NP_001128413.1 | |
Appetite-regulating hormone Protein | |
Research Use Only | |
GHRL | |
Recombinant Protein | |
E. coli | |
MPSPGTVCSL LLLGMLWLDL AMAGSSFLSP EHQRVQQRKE SKKPPAKLQP RALAGWLRPE DGGQAEGAED EMEVRFNAPF DVGIKLSGVQ YQQHSQALGK FLQDILWEEA KEAPADK | |
Greater than 80% as determined by SDS-PAGE. |
51738 | |
50 μg | |
Not determined. | |
Full Length of BC025791 | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |