missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Recombinant Human Apolipoprotein E/ApoE Protein
Shop All Bio Techne Products
Click to view available options
:
0.1mg; Unlabeled
Description
Recombinant bioactive protein containing 300 amino acids for Human Apolipoprotein E/ApoE The Recombinant Human Apolipoprotein E/ApoE Protein is derived from E. coli. The Recombinant Human Apolipoprotein E/ApoE Protein has been validated for the following applications: Western Blot, SDS-Page.
Specifications
Specifications
| Accession Number | NM_000041 |
| For Use With (Application) | SDS-PAGE |
| Formulation | Sterile filtered and lyophilized with no additives. |
| Gene ID (Entrez) | 348 |
| Molecular Weight (g/mol) | 34.3kDa |
| Name | Apolipoprotein E/ApoE protein |
| Purification Method | SDS-PAGE |
| Storage Requirements | −20°C. Avoid freeze-thaw cycles. |
| Cross Reactivity | Human |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction