missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Alkaline Phosphatase Protein
A DNA sequence encoding the Homo sapiens (Human) Alkaline Phosphatase, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8972-ye-50ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001622.2 | |
Alkaline Phosphatase Protein | |
Research Use Only | |
ALPI | |
Recombinant Protein | |
Yeast | |
VIPAEEENPA FWNRQAAEAL DAAKKLQPIQ KVAKNLILFL GDGLGVPTVT ATRILKGQKN GKLGPETPLA MDRFPYLALS KTYNVDRQVP DSAATATAYL CGVKANFQTI GLSAAARFNQ CNTTRGNEVI SVMNRAKQAG KSVGVVTTTR VQHASPAGTY AHTVNRNWYS DADMPASARQ EGCQDIATQL ISNMDIDVIL GGGRKYMFPM GTPDPEYPAD ASQNGIRLDG KNLVQEWLAK HQGAWYVWNR TELMQASLDQ SVTHLMGLFE PGDTKYEIHR DPTLDPSLME MTEAALRLLS RNPRGFYLFV EGGRIDHGHH EGVAYQALTE AVMFDDAIER AGQLTSEEDT LTLVTADHSH VFSFGGYTLR GSSIFGLAPS KAQDSKAYTS ILYGNGPGYV FNSGVRPDVN ESESGSPDYQ QQAAVPLSSE THGGEDVAVF ARGPQAHLVH GVQEQSFVAH VMAFAACLEP YTACDLAPPA CTTD | |
Greater than 90% as determined by SDS-PAGE. |
248 | |
50 μg | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |