missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Aldose 1-epimerase Protein

A DNA sequence encoding the Homo sapiens (Human) Aldose 1-epimerase, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7961-ec-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16051851

  • 2365.00 € / 1mg

Estimated Shipment: 22-11-2022
to see stock.
Add to basket

Description

Description

Specifications

Specifications

NP_620156.1
Aldose 1-epimerase Protein
1 mg
Not determined.
Full Length
Human
His-SUMO
Tris-based buffer, 50% glycerol
130589
Greater than 90% as determined by SDS-PAGE.
Research Use Only
GALM
Recombinant Protein
E. coli
ASVTRAVFGE LPSGGGTVEK FQLQSDLLRV DIISWGCTIT ALEVKDRQGR ASDVVLGFAE LEGYLQKQPY FGAVIGRVAN RIAKGTFKVD GKEYHLAINK EPNSLHGGVR GFDKVLWTPR VLSNGVQFSR ISPDGEEGYP GELKVWVTYT LDGGELIVNY RAQASQATPV NLTNHSYFNL AGQASPNIND HEVTIEADTY LPVDETLIPT GEVAPVQGTA FDLRKPVELG KHLQDFHLNG FDHNFCLKGS KEKHFCARVH HAASGRVLEV YTTQPGVQFY TGNFLDGTLK GKNGAVYPKH SGFCLETQNW PDAVNQPRFP PVLLRPGEEY DHTTWFKFSV A
Videos
SDS
Documents
Certificates
Promotions

Promotions