missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human 14-3-3 beta / YWHAB Protein

A DNA sequence encoding the Homo sapiens (Human) 14-3-3 beta / YWHAB, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP867-50ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16055881

  • 339.00 € / 50µg
Estimated Shipment: 14-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_003395.1
14-3-3 beta / YWHAB Protein
Research Use Only
YWHAB
Recombinant Protein
E. coli
MTMDKSELVQ KAKLAEQAER YDDMAAAMKA VTEQGHELSN EERNLLSVAY KNVVGARRSS WRVISSIEQK TERNEKKQQM GKEYREKIEA ELQDICNDVL ELLDKYLIPN ATQPESKVFY LKMKGDYFRY LSEVASGDNK QTTVSNSQQA YQEAFEISKK EMQPTHPIRL GLALNFSVFY YEILNSPEKA CSLAKTAFDE AIAELDTLNE ESYKDSTLIM QLLRDNLTLW TSENQGDEGD AGEGEN
Greater than 90% as determined by SDS-PAGE.
7529
50 μg
Not determined.
Full Length
Human
GST
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions