missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Guinea Pig IFNG / Interferon Gamma Protein
A DNA sequence encoding the Cavia porcellus (Guinea pig) IFNG / Interferon Gamma Protein, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8811-ec-1mg
Additional Details : Weight : 0.01000kg
Specifications
NP_001166345.1 | |
IFNG / Interferon Gamma Protein | |
Research Use Only | |
IFNG | |
Recombinant Protein | |
E. coli | |
QSRFTNEIRI LKNYFNADNS DVGDNGTLFV GILKNCQEES ERKIFQSQIV SFYFKLFEKH FTDNQTVQNS MNTIKEQIIT KFFKDNSSNK VQAFKNLIQI SVNDEHVQRQ AIIELKKVID DLSPNQRKRR RTQMLFQSRR ASK | |
Greater than 90% as determined by SDS-PAGE. |
100379568 | |
1 mg | |
Not determined. | |
Full Length | |
Guinea Pig | |
His | |
Tris-based buffer, 50% glycerol |