missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant E. coli Thioredoxin-2 Protein
A DNA sequence encoding the Escherichia coli (strain K12) Thioredoxin-2, was expressed in the hosts and tags indicated.
352.00 € - 4075.00 €
Specifications
Accession Number | NP_417077.1 |
---|---|
Gene ID (Entrez) | 947062 |
Name | Thioredoxin-2 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16087030
|
enQuireBio™
QP7012-YE-100UG |
100 μg |
1100.00 €
100µg |
Estimated Shipment: 25-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16007040
|
enQuireBio™
QP7012-YE-1MG |
1 mg |
4075.00 €
1mg |
Estimated Shipment: 25-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16097030
|
enQuireBio™
QP7012-YE-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16017040
|
enQuireBio™
QP7012-YE-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16027040
|
enQuireBio™
QP7012-YE-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16037040
|
enQuireBio™
QP7012-YE-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_417077.1 | |
Thioredoxin-2 Protein | |
Not determined. | |
Full Length | |
E. coli | |
His | |
Tris-based buffer, 50% glycerol |
947062 | |
Research Use Only | |
trxC | |
Recombinant Protein | |
Yeast | |
MNTVCTHCQA INRIPDDRIE DAAKCGRCGH DLFDGEVINA TGETLDKLLK DDLPVVIDFW APWCGPCRNF APIFEDVAQE RSGKVRFVKV NTEAERELSS RFGIRSIPTI MIFKNGQVVD MLNGAVPKAP FDSWLNESL | |
Greater than 90% as determined by SDS-PAGE. |