missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant E. coli nrdD Protein

A DNA sequence encoding the Escherichia coli (strain K12) nrdD, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP8848-ec-500ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16076811

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

Specifications

Specifications

NP_418659.1
nrdD Protein
Research Use Only
nrdD
Recombinant Protein
E. coli
MTPHVMKRDG CKVPFKSERI KEAILRAAKA AEVDDADYCA TVAAVVSEQM QGRNQVDINE IQTAVENQLM SGPYKQLARA YIEYRHDRDI EREKRGRLNQ EIRGLVEQTN ASLLNENANK DSKVIPTQRD LLAGIVAKHY ARQHLLPRDV VQAHERGDIH YHDLDYSPFF PMFNCMLIDL KGMLTQGFKM GNAEIEPPKS ISTATAVTAQ IIAQVASHIY GGTTINRIDE VLAPFVTASY NKHRKTAEEW NIPDAEGYAN SRTIKECYDA FQSLEYEVNT LHTANGQTPF VTFGFGLGTS WESRLIQESI LRNRIAGLGK NRKTAVFPKL VFAIRDGLNH KKGDPNYDIK QLALECASKR MYPDILNYDQ VVKVTGSFKT PMGCRSFLGV WENENGEQIH DGRNNLGVIS LNLPRIALEA KGDEATFWKL LDERLVLARK ALMTRIARLE GVKARVAPIL YMEGACGVRL NADDDVSEIF KNGRASISLG YIGIHETINA LFGGEHVYDN EQLRAKGIAI VERLRQAVDQ WKEETGYGFS LYSTPSENLC DRFCRLDTAE FGVVPGVTDK GYYTNSFHLD VEKKVNPYDK IDFEAPYPPL ANGGFICYGE YPNIQHNLKA LEDVWDYSYQ HVPYYGTNTP IDECYECGFT GEFECTSKGF TCPKCGNHDA SRVSVTRRVC GYLGSPDARP FNAGKQEEVK RRVKHLGNGQ IG
Greater than 90% as determined by SDS-PAGE.
948755
500 μg
Not determined.
Full Length
E. coli
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions