missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant E. coli nrdD Protein
A DNA sequence encoding the Escherichia coli (strain K12) nrdD, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8848-ec-200ug
Additional Details : Weight : 0.01000kg
Specifications
NP_418659.1 | |
nrdD Protein | |
Research Use Only | |
nrdD | |
Recombinant Protein | |
E. coli | |
MTPHVMKRDG CKVPFKSERI KEAILRAAKA AEVDDADYCA TVAAVVSEQM QGRNQVDINE IQTAVENQLM SGPYKQLARA YIEYRHDRDI EREKRGRLNQ EIRGLVEQTN ASLLNENANK DSKVIPTQRD LLAGIVAKHY ARQHLLPRDV VQAHERGDIH YHDLDYSPFF PMFNCMLIDL KGMLTQGFKM GNAEIEPPKS ISTATAVTAQ IIAQVASHIY GGTTINRIDE VLAPFVTASY NKHRKTAEEW NIPDAEGYAN SRTIKECYDA FQSLEYEVNT LHTANGQTPF VTFGFGLGTS WESRLIQESI LRNRIAGLGK NRKTAVFPKL VFAIRDGLNH KKGDPNYDIK QLALECASKR MYPDILNYDQ VVKVTGSFKT PMGCRSFLGV WENENGEQIH DGRNNLGVIS LNLPRIALEA KGDEATFWKL LDERLVLARK ALMTRIARLE GVKARVAPIL YMEGACGVRL NADDDVSEIF KNGRASISLG YIGIHETINA LFGGEHVYDN EQLRAKGIAI VERLRQAVDQ WKEETGYGFS LYSTPSENLC DRFCRLDTAE FGVVPGVTDK GYYTNSFHLD VEKKVNPYDK IDFEAPYPPL ANGGFICYGE YPNIQHNLKA LEDVWDYSYQ HVPYYGTNTP IDECYECGFT GEFECTSKGF TCPKCGNHDA SRVSVTRRVC GYLGSPDARP FNAGKQEEVK RRVKHLGNGQ IG | |
Greater than 90% as determined by SDS-PAGE. |
948755 | |
200 μg | |
Not determined. | |
Full Length | |
E. coli | |
His | |
Tris-based buffer, 50% glycerol |