missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant E. coli Glycerol kinase Protein

A DNA sequence encoding the Escherichia coli (strain K12) Glycerol kinase, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP8938-ec-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16057241

  • 985.00 € / 100µg
Estimated Shipment: 28-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_418361.1
Glycerol kinase Protein
Research Use Only
glpK
Recombinant Protein
E. coli
TEKKYIVALD QGTTSSRAVV MDHDANIISV SQREFEQIYP KPGWVEHDPM EIWATQSSTL VEVLAKADIS SDQIAAIGIT NQRETTIVWE KETGKPIYNA IVWQCRRTAE ICEHLKRDGL EDYIRSNTGL VIDPYFSGTK VKWILDHVEG SRERARRGEL LFGTVDTWLI WKMTQGRVHV TDYTNASRTM LFNIHTLDWD DKMLEVLDIP REMLPEVRRS SEVYGQTNIG GKGGTRIPIS GIAGDQQAAL FGQLCVKEGM AKNTYGTGCF MLMNTGEKAV KSENGLLTTI ACGPTGEVNY ALEGAVFMAG ASIQWLRDEM KLINDAYDSE YFATKVQNTN GVYVVPAFTG LGAPYWDPYA RGAIFGLTRG VNANHIIRAT LESIAYQTRD VLEAMQADSG IRLHALRVDG GAVANNFLMQ FQSDILGTRV ERPEVREVTA LGAAYLAGLA VGFWQNLDEL QEKAVIEREF RPGIETTERN YRYAGWKKAV KRAMAWEEHD E
Greater than 90% as determined by SDS-PAGE.
948423
100 μg
Not determined.
Full Length
E. coli
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions