missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant E. coli Glucose-6-phosphate 1-dehydrogenase Protein
A DNA sequence encoding the Escherichia coli (strain K12) Glucose-6-phosphate 1-dehydrogenase, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8937-ec-50ug
Additional Details : Weight : 0.01000kg
Specifications
NP_416366.1 | |
Glucose-6-phosphate 1-dehydrogenase Protein | |
Research Use Only | |
zwf | |
Recombinant Protein | |
E. coli | |
MAVTQTAQAC DLVIFGAKGD LARRKLLPSL YQLEKAGQLN PDTRIIGVGR ADWDKAAYTK VVREALETFM KETIDEGLWD TLSARLDFCN LDVNDTAAFS RLGAMLDQKN RITINYFAMP PSTFGAICKG LGEAKLNAKP ARVVMEKPLG TSLATSQEIN DQVGEYFEEC QVYRIDHYLG KETVLNLLAL RFANSLFVNN WDNRTIDHVE ITVAEEVGIE GRWGYFDKAG QMRDMIQNHL LQILCMIAMS PPSDLSADSI RDEKVKVLKS LRRIDRSNVR EKTVRGQYTA GFAQGKKVPG YLEEEGANKS SNTETFVAIR VDIDNWRWAG VPFYLRTGKR LPTKCSEVVV YFKTPELNLF KESWQDLPQN KLTIRLQPDE GVDIQVLNKV PGLDHKHNLQ ITKLDLSYSE TFNQTHLADA YERLLLETMR GIQALFVRRD EVEEAWKWVD SITEAWAMDN DAPKPYQAGT WGPVASVAMI TRDGRS | |
Greater than 90% as determined by SDS-PAGE. |
946370 | |
50 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
E. coli | |
GST | |
Tris-based buffer, 50% glycerol |