missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant E. Coli Chaperone Protein FIMC Protein

Product Code. 15997148
Change view
Click to view available options
Quantity:
1 mg
25 μg
5 μg
Unit Size:
1mg
25µg
5µg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15997148 5 μg 5µg
15987148 25 μg 25µg
15977148 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15997148 Supplier enQuireBio™ Supplier No. QP114025ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

A cDNA sequence encoding the Chaperone Protein FIMC was constructed and used to recombinantly synthesize the protein.

Specifications

Name Chaperone Protein FIMC Protein
Quantity 5 μg
Regulatory Status Research Use Only
Endotoxin Concentration Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Product Type Recombinant Protein
Cross Reactivity E. coli
Species E. coli
Protein Tag His
Sequence MGSSHHHHHH SSGLVPRGSH MGVALGATRV IYPAGQKQEQ LAVTNNDENS TYLIQSWVEN ADGVKDGRFI VTPPLFAMKG KKENTLRILD ATNNQLPQDR ESLFWMNVKA IPSMDKSKLT ENTLQLAIIS RIKLYYRPAK LALPPDQAAE KLRFRRSANS LTLINPTPYY LTVTELNAGT RVLENALVPP MGESTVKLPS DAGSNITYRT INDYGALTPK MTGVME
Buffer The FIMC solution (1 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 10% glycerol and 1mM DTT.
Purity or Quality Grade Greater than 95.0% as determined by SDS-PAGE.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.