missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant E. coli BirA His-tag Protein
Description
The pyrophosphate is subsequently hydrolyzed using Recombinant Yeast Inorganic Pyrophosphatase/PPA1 (ryPPA1) (Catalog # 8088-PP ). The specific activity is >10.0 pmol/min/μg, as measured under the described conditions.
Specifications
Specifications
| Accession Number | NP_418404.1 |
| Formulation | Supplied as a 0.2 μm filtered solution in MES, NaCl, TCEP, EDTA and Glycerol. |
| Gene ID (Entrez) | 948469 |
| Molecular Weight (g/mol) | Observed: 30-38 kDa, under reducing conditions |
| Quantity | 100 μg |
| Source | E. coli-derived e. coli BirA protein Lys2-Lys321, with an N-terminal Met & 6-His tag |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20°C to -70°C as supplied. 3 months, -20°C to -70°C under sterile conditions after opening. |
| Endotoxin Concentration | <0.10 EU / 1 μg of the protein by the LAL method. |
| Conjugate | Unconjugated |
| Purity or Quality Grade | >90%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie Blue Staining. |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?