missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant E. coli Beta-lactamase TEM Protein

Product Code. 16067880
Change view
Click to view available options
Quantity:
1 mg
10 μg
100 μg
200 μg
50 μg
500 μg
Unit Size:
100µg
10µg
1mg
200µg
500µg
50µg
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16067880 10 μg 10µg
16007890 50 μg 50µg
16057880 100 μg 100µg
16087880 200 μg 200µg
16097880 500 μg 500µg
16077880 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Product Code. 16067880 Supplier enQuireBio™ Supplier No. QP7256ec10ug

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

A DNA sequence encoding the Escherichia coli (strain K12) Beta-lactamase TEM, was expressed in the hosts and tags indicated.

Specifications

Accession Number NP_943295.1
Gene ID (Entrez) 10076131
Name Beta-lactamase TEM Protein
Quantity 10 μg
Regulatory Status Research Use Only
Endotoxin Concentration Not determined.
Gene Symbol bla
Protein Length Full Length
Product Type Recombinant Protein
Cross Reactivity E. coli
Species E. coli
Protein Tag His-SUMO
Sequence HPETLVKVKD AEDQLGARVG YIELDLNSGK ILESFRPEER FPMMSTFKVL LCGAVLSRVD AGQEQLGRRI HYSQNDLVEY SPVTEKHLTD GMTVRELCSA AITMSDNTAA NLLLTTIGGP KELTAFLHNM GDHVTRLDRW EPELNEAIPN DERDTTMPAA MATTLRKLLT GELLTLASRQ QLIDWMEADK VAGPLLRSAL PAGWFIADKS GAGERGSRGI IAALGPDGKP SRIVVIYTTG SQATMDERNR QIAEIGASLI KHW
Buffer Tris-based buffer, 50% glycerol
Purity or Quality Grade Greater than 90% as determined by SDS-PAGE.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.