missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant E. coli ATP-dependent DNA helicase Rep Protein

A DNA sequence encoding the Escherichia coli (strain K12) ATP-dependent DNA helicase Rep, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP8666-ec-200ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16095851

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

Specifications

Specifications

WP_001238899.1
ATP-dependent DNA helicase Rep Protein
Research Use Only
rep
Recombinant Protein
E. coli
MRLNPGQQQA VEFVTGPCLV LAGAGSGKTR VITNKIAHLI RGCGYQARHI AAVTFTNKAA REMKERVGQT LGRKEARGLM ISTFHTLGLD IIKREYAALG MKANFSLFDD TDQLALLKEL TEGLIEDDKV LLQQLISTIS NWKNDLKTPS QAAASAIGER DRIFAHCYGL YDAHLKACNV LDFDDLILLP TLLLQRNEEV RKRWQNKIRY LLVDEYQDTN TSQYELVKLL VGSRARFTVV GDDDQSIYSW RGARPQNLVL LSQDFPALKV IKLEQNYRSS GRILKAANIL IANNPHVFEK RLFSELGYGA ELKVLSANNE EHEAERVTGE LIAHHFVNKT QYKDYAILYR GNHQSRVFEK FLMQNRIPYK ISGGTSFFSR PEIKDLLAYL RVLTNPDDDS AFLRIVNTPK REIGPATLKK LGEWAMTRNK SMFTASFDMG LSQTLSGRGY EALTRFTHWL AEIQRLAERE PIAAVRDLIH GMDYESWLYE TSPSPKAAEM RMKNVNQLFS WMTEMLEGSE LDEPMTLTQV VTRFTLRDMM ERGESEEELD QVQLMTLHAS KGLEFPYVYM VGMEEGFLPH QSSIDEDNID EERRLAYVGI TRAQKELTFT LCKERRQYGE LVRPEPSRFL LELPQDDLIW EQERKVVSAE ERMQKGQSHL ANLKAMMAAK RGK
Greater than 90% as determined by SDS-PAGE.
948292
200 μg
Not determined.
Full Length
E. coli
GST
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions