Learn More
enQuireBio™ Recombinant E. coli 30S ribosomal protein S8 Protein
Product Code. p-7155263
Click to view available options
Quantity:
1 mg
10 μg
100 μg
200 μg
50 μg
500 μg
Unit Size:
100µg
10µg
1mg
200µg
500µg
50µg
Product Code. 16065551
Brand: enQuireBio™ QP8610ec10ug
Please to purchase this item. Need a web account? Register with us today!
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
A DNA sequence encoding the Escherichia coli (strain K12) 30S ribosomal protein S8, was expressed in the hosts and tags indicated.
Specifications
Accession Number NP_417765.1
Gene ID (Entrez) 31660798
Name 30S ribosomal protein S8 Protein
Quantity 10 μg
Regulatory Status Research Use Only
Endotoxin Concentration Not determined.
Gene Symbol rpsH
Protein Length Full Length
Product Type Recombinant Protein
Cross Reactivity E. coli
Species E. coli
Protein Tag His
Sequence SMQDPIADML TRIRNGQAAN KAAVTMPSSK LKVAIANVLK EEGFIEDFKV EGDTKPELEL TLKYFQGKAV VESIQRVSRP GLRIYKRKDE LPKVMAGLGI AVVSTSKGVM TDRAARQAGL GGEIICYVA
Buffer Tris-based buffer, 50% glycerol
Purity or Quality Grade Greater than 90% as determined by SDS-PAGE.