missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Dengue Virus Dengue Envelope-4 22kDa Protein

Product Code. 15907878
Change view
Click to view available options
Quantity:
1 mg
100 μg
500 μg
Unit Size:
100µg
1mg
500µg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15907878 1 mg 1mg
15997868 100 μg 100µg
15917878 500 μg 500µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15907878 Supplier enQuireBio™ Supplier No. QP116361mg
  • ONLINE EXCLUSIVE PRICE
    1905.75 €
    1558.00 € / 1mg
    Save 347.75 €
    18% Off
 Restricted Item
Estimated Shipment: 21-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A cDNA sequence encoding the Dengue Envelope-4 22kDa was constructed and used to recombinantly synthesize the protein.

Specifications

Name Dengue Envelope-4 22kDa Protein
Quantity 1 mg
Regulatory Status RUO
Endotoxin Concentration Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Product Type Recombinant Protein
Cross Reactivity Dengue Virus
Species E. coli
Protein Tag Untagged
Buffer Phosphate buffered saline, pH 7.4.
Purity or Quality Grade Protein is >95% pure as determined by 12% PAGE (coomassie staining).
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.