Learn More
enQuireBio™ Recombinant Cytomegalovirus Uncharacterized protein UL131 Protein
Product Code. 16077340
Click to view available options
Quantity:
1 mg
10 μg
100 μg
200 μg
50 μg
500 μg
Unit Size:
100µg
10µg
1mg
200µg
500µg
50µg
Product Code. 16077340
Brand: enQuireBio™ QP7086iv100ug
Please to purchase this item. Need a web account? Register with us today!
A DNA sequence encoding the Human cytomegalovirus (strain AD169) (HHV-5) (Human herpesvirus 5) Uncharacterized protein UL131, was expressed in the hosts and tags indicated.
Specifications
Name Uncharacterized protein UL131 Protein
Quantity 100 μg
Regulatory Status Research Use Only
Endotoxin Concentration Not determined.
Gene Symbol UL131
Protein Length Full Length
Product Type Recombinant Protein
Cross Reactivity Cytomegalovirus
Species E. coli
Protein Tag His-SUMO
Sequence MCMMSHNKAF FLSLQHAAVS GVAVCLSVRR GAGSVPAGNR GKKTIITEYR ITGTRALARC PTKPVTSMWN SSWTSR
Buffer Tris-based buffer, 50% glycerol
Purity or Quality Grade Greater than 80% as determined by SDS-PAGE.