missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Cytomegalovirus Envelope glycoprotein 1

A DNA sequence encoding the Human cytomegalovirus Envelope glycoprotein L, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7456-ye-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16008910

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

Specifications

Specifications

YP_081555.1
Envelope glycoprotein 1
Research Use Only
gL
Recombinant Protein
Yeast
AAVSVAPTAA EKVPAECPEL TRRCLLGEVF EGDKYESWLR PLVNVTGRDG PLSQLIRYRP VTPEAANSVL LDEAFLDTLA LLYNNPDQLR ALLTLLSSDT APRWMTVMRG YSECGDGSPA VYTCVDDLCR GYDLTRLSYG RSIFTEHVLG FELVPPSLFN VVVAIRNEAT RTNRAVRLPV STAAAPEGIT LFYGLYNAVK EFCLRHQLDP PLLRHLDKYY AGLPPELKQT RVNLPAHSRY GPQAVDAR
Greater than 90% as determined by SDS-PAGE.
3077416
1 mg
Not determined.
Full Length
Cytomegalovirus
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions