missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Cynomolgus Monkey Ly6E Fc Chimera Protein
Description
Measured by its ability to inhibit anti-CD3 antibody induced IL-2 or IFN-gamma secretion by human T cells. The ED50 for this effect 1-10 μg/mL.
Specifications
Specifications
| Accession Number | NP_001270930.1 |
| For Use With (Application) | Bioactivity |
| Formulation | Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. |
| Gene ID (Entrez) | 4061 |
| Molecular Weight (g/mol) | M.W.-observed: 38-50 kDa, under reducing conditions, M.W.-theoretical: 35 kDa |
| Quantity | 50 μg |
| Source | Chinese Hamster Ovary cell line, CHO-derived cynomolgus monkey Ly6E protein Cynomolgus Monkey Ly6E (Leu21-Ser101) (N-terminus) Accession # NP_001270930.1 3 x GSL Human IgG1 (Pro100-Lys330) (C-terminus) |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution. |
| Endotoxin Concentration | <1.0 EU / 1 μg of the protein by the LAL method. |
| Gene Alias | Ly-6 E, lymphocyte antigen 6 complex, locus E, Retinoic acid-induced gene E protein, RIGE, RIGE9804, RIG-Ely-6 E, SCA2, SCA2Ly-6 E, SCA-2 lymphocyte antigen 6 E, Stem cell antigen 2, Thymic shared antigen 1, TSA1, TSA-1 retinoic acid induced gene E |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?