missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Bovine Interferon tau-1 Protein

A DNA sequence encoding the Bos taurus (Bovine) Interferon tau-1, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7076-ye-500ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16097300

  • 2650.00 € / 500µg
Estimated Shipment: 25-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_001015511.3
Interferon tau-1 Protein
Research Use Only
IFNT1
Recombinant Protein
Yeast
CYLSEDHMLG ARENLRLLAR MNRLSPHPCL QDRKDFGLPQ EMVEGNQLQK DQAISVLHEM LQQCFNLFYT EHSSAAWNTT LLEQLCTGLQ QQLEDLDACL GPVMGEKDSD MGRMGPILTV KKYFQGIHVY LKEKEYSDCA WEIIRVEMMR ALSSSTTLQK RLRKMGGDLN SL
Greater than 90% as determined by SDS-PAGE.
317698
500 μg
Not determined.
Full Length
Bovine
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions