missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Bovine Complement C3 Protein
A DNA sequence encoding the Bos taurus (Bovine) Complement C3, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7640-ec-1mg
Additional Details : Weight : 0.01000kg
Specifications
NP_001035559.2 | |
Complement C3 Protein | |
Research Use Only | |
C3 | |
Recombinant Protein | |
E. coli | |
SDLDDDIIPE EDIISRSQFP ESWLWTVIED LKQADKNGIS TKLMNVFLKD SITTWEILAV SLSDKKGICV ADPYEVTVMQ DFFIDLRLPY SVVRNEQVEI RAILYNYREA ENLKVRVELL YNPAFCSLAT AKKRHQQTIT IPARSSVAVP YVIVPLKIGL HEVEVKAAVY NHFISDGVKK TLKVVPEGVR VNKTVAVRTL NPEHLGQ | |
Greater than 90% as determined by SDS-PAGE. |
280677 | |
1 mg | |
Not determined. | |
Partial (see sequence information for more details). | |
Bovine | |
His-SUMO | |
Tris-based buffer, 50% glycerol |