missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Bacteriophage Recombination protein uvsY Protein
A DNA sequence encoding the Enterobacteria phage T4 (Bacteriophage T4) Recombination protein uvsY, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7417-ec-50ug
Additional Details : Weight : 0.01000kg
Specifications
NP_049799.2 | |
Recombination protein uvsY Protein | |
Research Use Only | |
uvsY | |
Recombinant Protein | |
E. coli | |
MRLEDLQEEL KKDVFIDSTK LQYEAANNVM LYSKWLNKHS SIKKEMLRIE AQKKVALKAR LDYYSGRGDG DEFSMDRYEK SEMKTVLSAD KDVLKVDTSL QYWGILLDFC SGALDAIKSR GFAIKHIQDM RAFEAGK | |
Greater than 86.7% as determined by SDS-PAGE. |
1258547 | |
50 μg | |
Not determined. | |
Full Length | |
Bacteriophage | |
His-SUMO | |
Tris-based buffer, 50% glycerol |