Learn More
Rebastinib, MedChemExpress
Product Code. 16405498
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
Product Code. 16405498
Brand: Medchem Express HY13024
Please to purchase this item. Need a web account? Register with us today!
MedChemExpress Rebastinib (DCC-2036) is an orally active, non-ATP-competitive Bcr-Abl inhibitor for Abl1WT and Abl1T315I with IC50s of 0.8 nM and 4 nM, respectively. Rebastinib also inhibits SRC, KDR, FLT3, and Tie-2, and has low activity to seen towards c-Kit.
Rebastinib (DCC-2036) is an orally active, non-ATP-competitive Bcr-Abl inhibitor for Abl1WT and Abl1T315I with IC50s of 0.8 nM and 4 nM, respectively. Rebastinib also inhibits SRC, KDR, FLT3, and Tie-2, and has low activity to seen towards c-Kit.
Specifications
Chemical Name or Material Rebastinib
CAS 1020172-07-9
Color Off-White
Purity Grade Notes Research
Molecular Formula C30H28FN7O3
Quantity 10 mg
Synonym DCC-2036
Solubility Information DMSO : 50 mg/mL (90.32 mM; ultrasonic and warming and heat to 80°C)
SMILES CC(C)(C)C1=NN(C(NC(NC2=C(F)C=C(OC3=CC(C(NC)=O)=NC=C3)C=C2)=O)=C1)C4=CC=C5C(C=CC=N5)=C4
Molecular Weight (g/mol) 553.59
Formula Weight 553.59
Percent Purity 98.0%
Grade Research
Physical Form Powder
For Use With (Application) Cancer-Kinase/protease