Learn More
behr Labor-Technik™ Reaction Vessel for COD Determination
Product Code. 10085100
Change view
Click to view available options
Volume (Metric):
175 mL
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) unitSize
10085100 175 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10085100 Supplier behr Labor-Technik™ Supplier No. B00217863
  • ONLINE EXCLUSIVE PRICE
    109.20 €
    82.16 € / Pack of 1
    Save 27.04 €
    24% Off
 Restricted Item
Estimated Shipment: 13-08-2026
Add to basket
Add to basket
Use these vessels for COD Determination. behr Labor-Technik™ Reaction Vessels feature 50 and 100mL scale marks for easy filling prior to titration.
  • 175mL Reaction vessel with vessel adapters
  • NS 29 and 100mL scale mark
  • Flat bottom for improved heat transfer
  • PVDF collar for insulation purposes
  • Gentle transport in EG rack insert
Specifications
Certifications/Compliance DIN/DEV
Volume (Metric) 175 mL
Type Reaction Vessel