missing translation for 'onlineSavingsMsg'
Learn More

RDH13 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Product Code. 16855544 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
20 μL
150 μL
Unit Size:
150µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16855544 20 μL 20µL
16845544 150 μL 150µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16855544 Supplier Proteintech Supplier No. 160671AP20UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Does not exhibit retinol dehydrogenase (RDH) activity in vitro.

Specifications

Antigen RDH13
Applications Western Blot
Classification Polyclonal
Concentration 0.15 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene RDH13
Gene Accession No. Q8CEE7, Q8NBN7
Gene Alias RDH13, Retinol dehydrogenase 13, SDR7C3, UNQ736/PRO1430
Gene Symbols Rdh13
Host Species Rabbit
Immunogen RDH13 Fusion Protein Ag8944
Purification Method Antigen Affinity Chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 108841, 112724, 361504
Target Species Mouse, Rat, Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.