Learn More
KNF™ RC 600 Rotary Evaporators
The RC 600 Rotary Evaporator provides a user-friendly interface while saving valuable lab bench space with its small footprint. The electric lift allows for easy and safe operation. Additional benefits are the simple flask exchange or the cordless heating bath.
Brand:  KNF™ 302500/302686
Additional Details : Weight : 9.10000kg
Product Code. 15583870
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Description
Description
  • Shaped bath uses less fluid and energy
  • Small footprint to save bench/fumehood space
  • Easy fit, one handed flask attachment
  • Cordless bath with spout and hand-holds for safety
  • Simple intuitive controls for ease of use
  • Memory function of flask height and rotation speed
  • Optional 'intelligent' protective bath cover, can stop rotation and lift flask from bath if left open during operation
  • Safe and easy adjustment of flask angle without danger of dropping condenser
  • High efficiency condenser with anti 'chimney effect' feature
  • Quiet operation
  • Automatic vacuum control if used with KNF SC 920 G Vacuum System
  • Dry ice condenser available
Model Specific:
Model: RC600
  • Fixed to tower for more usable bench space
Model: RC900
  • Wireless remote control allows full operation from outside closed fume cupboard for safety and energy saving
  • Tubing hidden inside for more usable bench space
  • Adjustable alternating flask rotation for powder drying
Specifications
Specifications
Plug type: EU
With bath cover
RC600
LCD
Rotary Evaporator