Learn More
KNF™ RC 600 Rotary Evaporators
The RC 600 Rotary Evaporator provides a user-friendly interface while saving valuable lab bench space with its small footprint. The electric lift allows for easy and safe operation. Additional benefits are the simple flask exchange or the cordless heating bath.
Brand: KNF™ 126001/300576
Additional Details : Weight : 9.60000kg
Description
- Shaped bath uses less fluid and energy
- Small footprint to save bench/fumehood space
- Easy fit, one handed flask attachment
- Cordless bath with spout and hand-holds for safety
- Simple intuitive controls for ease of use
- Memory function of flask height and rotation speed
- Optional 'intelligent' protective bath cover, can stop rotation and lift flask from bath if left open during operation
- Safe and easy adjustment of flask angle without danger of dropping condenser
- High efficiency condenser with anti 'chimney effect' feature
- Quiet operation
- Automatic vacuum control if used with KNF SC 920 G Vacuum System
- Dry ice condenser available
- Fixed to tower for more usable bench space
- Wireless remote control allows full operation from outside closed fume cupboard for safety and energy saving
- Tubing hidden inside for more usable bench space
- Adjustable alternating flask rotation for powder drying
Specifications
Plug type: EU | |
With bath cover | |
RC900 |
25 to 250 rpm | |
Rotary Evaporator |