missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RBM7 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA541862
This item is not returnable.
View return policy
Description
Peptide sequence: SFNQSSSSQW RQGTPSSQRK VRMNSYPYLA DRHYSREQRY TDHGSDHHYR Sequence homology: Cow: 79%; Dog: 93%; Guinea Pig: 86%; Human: 100%; Mouse: 92%; Pig: 93%; Rabbit: 85%; Rat: 100%.
RBM7 contains 1 RRM (RNA recognition motif) domain. It is possible involved in germ cell RNA processing and meiosis.
Specifications
| RBM7 | |
| Polyclonal | |
| Unconjugated | |
| RBM7 | |
| 1200007M24Rik; 1500011D06Rik; AU041934; AW554393; RBM7; RNA binding motif protein 7; rna-binding; RNA-binding motif protein 7; RNA-binding protein 7 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 10179 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q9Y580 | |
| RBM7 | |
| Synthetic peptide directed towards the middle region of human RBM7. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction