missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ RBM34 Polyclonal Antibody
GREENER_CHOICE

Product Code. 16673945
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16673945 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16673945 Supplier Invitrogen™ Supplier No. PA599701

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total RBM34.

RNA binding motif protein 34 (RBM34) is an RRM (RNA recognition motif)-containing protein. Alternative names for RBM34 Antibody include KIAA0117 antibody, RNA-binding protein 34 antibody, RNA-binding motif protein 34 antibody, RNA binding motif protein 34 antibody.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RBM34
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene RBM34
Gene Accession No. P42696, Q8C5L7
Gene Alias 4930547K05Rik; 6330444B11; D8Ertd233e; KIAA0117; Rbm34; RGD1310161; RNA binding motif protein 34; RNA-binding motif protein 34; RNA-binding protein 34
Gene Symbols RBM34
Host Species Rabbit
Immunogen A synthesized peptide derived from human RBM34(Accession P42696), corresponding to amino acid residues A344-A394.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23029, 52202
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.