missing translation for 'onlineSavingsMsg'
Learn More

Rbin-1, MedChemExpress

Product Code. 16414466
Encompass_Preferred
Click to view available options
Quantity:
1 mg
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
1mg
25mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16414466

Brand: Medchem Express HY100816

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Rbin-1 is a potent, reversible, and specific chemical inhibitor of eukaryotic ribosome biogenesis. Rbin-1 inhibits the ATPase with GI50 of 136 nM. Rbin-1 is a potent and selective chemical inhibitor of Midasin (Mdn1).

Rbin-1 is a potent, reversible, and specific chemical inhibitor of eukaryotic ribosome biogenesis. Rbin-1 inhibits the ATPase with GI50 of 136 nM. Rbin-1 is a potent and selective chemical inhibitor of Midasin (Mdn1).

Specifications

Chemical Name or Material Rbin-1
Physical Form Solid
CAS 328023-11-6
Color Yellow
Quantity 100 mg
Purity Grade Notes Research
Molecular Formula C13H12N4S
For Use With (Application) Metabolism-sugar/lipid metabolism
Synonym Ribozinoindole-1
Solubility Information DMSO : ≥ 31 mg/mL (120.94 mM)
SMILES C=C(C)CSC1=NN=C2C(NC3=C2C=CC=C3)=N1
Molecular Weight (g/mol) 256.33
Formula Weight 256.33
Percent Purity 99.33%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.